Order Entry
Export
Orders LinkContactUsLinkComponent
Fluoride ISE standard solutions

Fluoride ISE standard solutions

Fluoride standard solution, 1,00 µg/ml, ±0,02 µg/ml, H₂O, Glass bottle, 500 mL
Catalog # HACH29149
Supplier:   Hach
Fluoride ISE standard solutions
Catalog # HACH29149
Supplier:  Hach
CAS Number:  
Fluoride ISE standard solutions

Fluoride standard solution, 1,00 µg/ml, ±0,02 µg/ml, H₂O, Glass bottle, 500 mL

CAS Number:  
Price
41,10 €
Log in for availability
1 * 500 mL
Product Details & Documents
  • Traceable to NIST standard reference materials
  • Designed for use with Hach meters portfolio and other meters
  • Reliable packaging

Caution: 1 µg/ml = 1 mg/l

Documents

Specifications

Specifications

  • Concentration:
    1,00 µg/ml
  • Matrix:
    H₂O
  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Description:
    Fluoride standard solution
  • Accuracy:
    ±0,02 µg/ml

Specifications