Order Entry
Export
Orders LinkContactUsLinkComponent
Lead Standard Solutions, NIST

Lead Standard Solutions, NIST

Lead, Pb, 100 mg/L, 100 mL
Catalog # 705-0051
Supplier:   Hach
Lead Standard Solutions, NIST
Catalog # 705-0051
Supplier:  Hach
CAS Number:  
Lead Standard Solutions, NIST
Lead Standard Solutions, NIST

Lead, Pb, 100 mg/L, 100 mL

CAS Number:  
Price
50,50 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solutions for accuracy checks in lead determinations.

Specifications

Specifications

  • Concentration:
    100 mg/L
  • Pk:
    100 mL
  • Element:
    Lead
  • Symbol:
    Pb

Specifications