Orders LinkContactUsLinkComponent
Visual comparator test kits, silica
Visual comparator test kits, silica
Silica LR, SI-7, 0 - 1,0 mg/l, 100, 1 KIT
Catalog # HACH2255000
Supplier:   Hach
Visual comparator test kits, silica
Catalog # HACH2255000
Supplier:  Hach
CAS Number:  
Visual comparator test kits, silica
Visual comparator test kits, silica
Silica LR, SI-7, 0 - 1,0 mg/l, 100, 1 KIT
CAS Number:  
Price
313,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Silica LR, SI-7
  • Tests per kit:
    100
  • Range:
    0 - 1,0 mg/l
Specifications