Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0,1 - 0,50 mg/l NO₃- N, Cadmium reduction, All except DR3000, DR2010, DR2000, PC II, 10 ml, PP, 100, 100
705-0132
: Hach
For determination of nitrogen-nitrate by the cadmium reduction method.
- Chemical method:Cadmium reduction
- Pk:100
- Description:Nitrogen nitrate reagent set
- Sample size:10 ml
- Type:PP
- For unit:All except DR3000, DR2010, DR2000, PC II
- Tests per kit:100
- Range:0,1 - 0,50 mg/l NO₃- N