Caps for bottles, open top, Microlink®, WHEATON®
Screw cap, solid top, with PP/silicone/PTFE liner, 33-430, Black
Catalog # WHEA240106
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
Screw caps are ideal for use with biological media, volatile solvents, corrosive chemicals and sensitive compounds.
- PTFE or PP on the exposed face of the liner imparts an inert, chemical-resistant surface
- Open top caps provide access with a syringe
Ultrasonically welded liner to the cap eliminates the possibility of glue contamination and bond failure on autoclaving. Low-compression-set silicone provides prolonged period of time sample storage over wide temperature ranges.
Specifications
- Thread:33-430
- Colour:Black
- Description:Screw cap, solid top, with PP/silicone/PTFE liner
- Seq:11