Orders LinkContactUsLinkComponent
Post-mortem scalpel handles
PM40 epoxy powder coated, Blade no's PM40 and PM40B, 220 mm
Catalog # 233-0116
Supplier:   SWANN MORTON
Post-mortem scalpel handles
Catalog # 233-0116
Supplier:  SWANN MORTON
Post-mortem scalpel handles
Post-mortem scalpel handles
PM40 epoxy powder coated, Blade no's PM40 and PM40B, 220 mm
Price
29,70 €
Log in for availability
1 * 1 items
Product Details & Documents
A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
  • PM40 range with traditional screw fitting
  • PM60 range with standard blade fitting
PM40B and PM60B blades are 'bull nose' blades for added safety and protection against accidental sharps injuries.
 : The PM8 and PM40 stainless steel handles and all blades are supplied individually wrapped.
Specifications
  • For:
    Blade no's PM40 and PM40B
  • Type:
    PM40 epoxy powder coated
  • Length:
    220 mm
  • Seq:
    03