Polysorbate 80, GPR RECTAPUR®
Plastic bottle, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
- CAS Number:9005-65-6
- MDL Number:MFCD00082107
- Merck Index:13,07664
- Boiling Pt:>100 °C (1013 hPa)
- Melting Pt:–21 °C
- Density:1.062 g/cm³ (20 °C)
- Flash Pt:149 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:500-019-9
Specifications
Specification Test Results
- Identification:Passes test
- Density (20/4):1.070 - 1.090
- Hydroxyl value:65 - 80
- Saponification value:45 - 55
- Acid value:Max 2 mg KOH/g
- Water content:Max 3%
Specifications
- Pack type:Plastic bottle
- Pk:1 L



