Order Entry
Export
Orders LinkContactUsLinkComponent
D-(+)-Sucrose, GPR RECTAPUR®

D-(+)-Sucrose, GPR RECTAPUR®

Plastic bucket, 25 kg
Catalog # 27478.467
Supplier:   VWR Chemicals
D-(+)-Sucrose, GPR RECTAPUR®
Catalog # 27478.467
Supplier:  VWR Chemicals
CAS Number:  57-50-1
D-(+)-Sucrose, GPR RECTAPUR®
D-(+)-Sucrose, GPR RECTAPUR®

Plastic bucket, 25 kg

CAS Number:  57-50-1
Price
395,00 €
Log in for availability
1 * 25 kg
Product Details & Documents
Synonyms:  β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
  • CAS Number:57-50-1
  • Formula:C₁₂H₂₂O₁₁
  • MDL Number:MFCD00006626
  • Molecular Weight:342.3 g/mol
  • Merck Index:13,08966
  • Melting Pt:169…170 °C
  • Density:1.29 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-334-9
Specifications

Specification Test Results

  • Specific optical rotation (2 %; water):
    66 - 67 °
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Ignition residue (SO4):
    Max. 0.2 %

Specifications

  • Pack type:
    Plastic bucket
  • Pk:
    25 kg

Specifications