Sand Fontainebleau, TECHNICAL
Plastic bucket, 5 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
- CAS Number:7631-86-9
- Formula:SiO₂
- Molecular Weight:60.08 g/mol
- Boiling Pt:2230 °C (1013 hPa)
- Storage Temperature:Ambient
- EINECS:238-878-4
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000) 33L Plastic bucket UN Z (ZPACK13247037) 5L Plastic bucket UN Z (ZPACK13247005)
Packaging Information
P20_VWRC27460.295_SPEC_EN.pdf P20_VWRC27460.364_SPEC_EN.pdf P20_VWRC27460.460_SPEC_EN.pdf spec_VWRC27460.295 spec_VWRC27460.295
Product specification
More
VWR® for Drying and Filtration
Product Literature
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bucket
- Pk:5 kg



