0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Sand Fontainebleau
Sand Fontainebleau, TECHNICAL
Sand Fontainebleau, TECHNICAL
Sand Fontainebleau, TECHNICAL
Plastic bottle for solids, 1 kg
27460.295
:
VWR Chemicals
Sand Fontainebleau, TECHNICAL
27460.295
:
VWR Chemicals
:
7631-86-9
Expand image
Sand Fontainebleau, TECHNICAL
Plastic bottle for solids, 1 kg
Add to cart
:
7631-86-9
37,40 €
Unit of Measure
1 * 1 kg
-
+
Add to cart
:
7631-86-9
:
SiO₂
:
60.08 g/mol
:
2230 °C (1013 hPa)
:
Ambient
:
238-878-4
1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
33L Plastic bucket UN Z (ZPACK13247037)
5L Plastic bucket UN Z (ZPACK13247005)
P20_VWRC27460.295_SPEC_EN.pdf
P20_VWRC27460.364_SPEC_EN.pdf
P20_VWRC27460.460_SPEC_EN.pdf
spec_VWRC27460.295
spec_VWRC27460.295
SDS_EN_UK_VWRC27460.pdf
sds_VWRC27460
VWR® for Drying and Filtration
Identification:
Passes test
Pack type:
Plastic bottle for solids
Pk:
1 kg