Order Entry
Export
Orders LinkContactUsLinkComponent
Dichloromethane ≥98% stabilised, TECHNICAL

Dichloromethane ≥98% stabilised, TECHNICAL

Glass bottle, 1 L
Catalog # 25629.295
Health
Supplier:   VWR Chemicals
Dichloromethane ≥98% stabilised, TECHNICAL
Catalog # 25629.295
Supplier:  VWR Chemicals
CAS Number:  75-09-2
Dichloromethane ≥98% stabilised, TECHNICAL
Dichloromethane ≥98% stabilised, TECHNICAL

Glass bottle, 1 L

CAS Number:  75-09-2
Price
36,30 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39.8 °C (1013 hPa)
  • Melting Pt:–95 °C
  • Density:1.322 g/cm³ (20 °C)
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilised with ethanol 0.15 %
  • Storage Temperature:Ambient
  • UN:1593
  • REACH:01-2119480404-41
Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 98 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications