Order Entry
Export
Orders LinkContactUsLinkComponent
Rubber washers

Rubber washers

Washer, I.Ø×H 40×7 mm, 100 mm
  DENE2404010
Rubber washers
  DENE2404010
Rubber washers
Rubber washers

Washer, I.Ø×H 40×7 mm, 100 mm

 
15,50 €
Log in for availability
1 * 1 items
 

These washers are made from red natural rubber.

  • Hardness 40 Shore A
Specifications
  • Ø:
    100 mm
  • Description:
    Washer, I.Ø×H 40×7 mm
  • Seq:
    04