Rubber washers
Washer, I.Ø×H 25×7 mm, 70 mm
DENE2402570
Jump to:
These washers are made from red natural rubber.
- Hardness 40 Shore A
Specifications
- Ø:70 mm
- Description:Washer, I.Ø×H 25×7 mm
- Seq:02
These washers are made from red natural rubber.