Paraffin, liquid, TECHNICAL
Plastic bottle, 1 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Wax Paraffin, Paraffin wax
- CAS Number:8042-47-5
- Merck Index:13,07093
- Boiling Pt:300-500 °C (1013 hPa)
- Density:0.872 g/cm³ (20 °C)
- Flash Pt:215 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:232-384-2
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Packaging Information
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bottle
- Pk:1 L



