Ethyl acetate ≥99%, GPR RECTAPUR®
Plastic bottle, 2,5 L
Supplier: VWR Chemicals
Ethyl acetate ≥99%, GPR RECTAPUR®
Catalog # 23880.324
Supplier: VWR Chemicals
CAS Number: 141-78-6
Jump to:
Product Details & Documents
Synonyms: Acetic acid ethyl ester
- CAS Number:141-78-6
- Formula:CH₃COOC₂H₅
- MDL Number:MFCD00009171
- Molecular Weight:88.11 g/mol
- Merck Index:13,03792
- Boiling Pt:77.1 °C (1013 hPa)
- Melting Pt:–83 °C
- Density:0.902 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Storage Temperature:Ambient
- EINECS:205-500-4
- UN:1173
- ADR:3,II
- REACH:01-2119475103-46
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Plastic bottle for liquids DIN 45 UN X (ZPACK13962500) 225L Metal drum UN (ZPACE8010050) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic jerrycan for liquids DIN 42 (ZPACK13951025)
Specifications
Specification Test Results
- Assay:Min. 99 %
- IR Spectrum:Passes test
- Acidity:Max. 0.008 meq/g
- Boiling point:76.0 - 77.5 °C
- Density (20/4):0.899 - 0.901
- Refractive index (20°C):1.371 - 1.373
- Substances coloured by H2SO4:Max. 60 APHA
- Ethanol:Max. 0.5 %
- Evaporation residue:Max. 10 ppm
- Heavy metals (as Pb):Max. 2 ppm
- Water:Max. 0.05 %
Specifications
- Pack type:Plastic bottle
- Pk:2,5 L
- Cat. No.:23880.324
- Environmental certification:Chem21 Solvent Selection Guide recommended by default and after discussion
- Environmentally Preferable:Safer Human and Environmental Health



