Order Entry
Export
Orders LinkContactUsLinkComponent
 

Diethyl ether stabilised Ph. Eur.

Steel drum, 25 L
Catalog # 23819.460
Flammable
Irritant
Supplier:   VWR Chemicals
Diethyl ether stabilised Ph. Eur.
Catalog # 23819.460
Supplier:  VWR Chemicals
CAS Number:  60-29-7
Diethyl ether stabilised Ph. Eur.
Diethyl ether stabilised Ph. Eur.

Steel drum, 25 L

CAS Number:  60-29-7
Price
788,00 €
Log in for availability
1 * 25 L
Product Details & Documents

Appearance: Clear, colourless, volatile, very mobile liquid.

 : Not for use as Active Pharmaceutical Ingredient (API)

Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specification Test Results

  • Assay (Ionol/BHT):
    Passes test
  • Appearance:
    Conforms (see CoA/CoS)
  • Acidity:
    Passes test
  • Relative density:
    0.714 - 0.716
  • Distillation range:
    34.0 - 35.0 °C
  • Acetone and aldehydes:
    Passes test
  • Peroxides:
    Passes test
  • Non-volatile matter:
    ≤ 20 mg/l
  • Unspecified impurities:
    ≤ 0.05 %
  • Total impurities:
    ≤ 0.1 %
  • Reporting threshold:
    ≤ 0.03 %
  • Water:
    ≤ 2 g/l
  • Residual solvents:
    Passes test
  • As (Arsenic):
    ≤ 0.1 ppm
  • Cd (Cadmium):
    ≤ 0.1 ppm
  • Co (Cobalt):
    ≤ 0.1 ppm
  • Hg (Mercury):
    ≤ 0.1 ppm
  • Ni (Nickel):
    ≤ 0.1 ppm
  • Pb (Lead):
    ≤ 0.1 ppm
  • V (Vanadium):
    ≤ 0.1 ppm
  • Conforms to Ph.Eur.:
    Passes test

Specifications

  • Pack type:
    Steel drum
  • Pk:
    25 L

Specifications