Order Entry
Export
Orders LinkContactUsLinkComponent
Diethyl ether ≥99% stabilised, GPR RECTAPUR®

Diethyl ether ≥99% stabilised, GPR RECTAPUR®

Aluminium bottle, 5 L
Catalog # 23809.363
Flammable
Irritant
Supplier:   VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Catalog # 23809.363
Supplier:  VWR Chemicals
CAS Number:  60-29-7
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
Diethyl ether ≥99% stabilised, GPR RECTAPUR®

Aluminium bottle, 5 L

CAS Number:  60-29-7
Price
183,00 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
  • CAS Number:60-29-7
  • Formula:(CH₃CH₂)₂O
  • MDL Number:MFCD00011646
  • Molecular Weight:74.12 g/mol
  • Merck Index:12,03852
  • Boiling Pt:35 °C (1013 hPa)
  • Melting Pt:–123 °C
  • Density:0.7135 g/cm³ (20 °C)
  • Flash Pt:–40 °C
  • UN:1155
  • ADR:3,I
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 4-7 ppm
  • Storage Temperature:Ambient
  • EINECS:200-467-2
  • REACH:01-2119535785-29
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Assay (Ionol/BHT):
    4 - 10 ppm
  • Acidity:
    Max. 0.0003 meq/g
  • Evaporation residue:
    Max. 20 ppm
  • Water:
    Max. 0.2 %

Specifications

  • Pack type:
    Aluminium bottle
  • Pk:
    5 L

Specifications