Order Entry
Export
Orders LinkContactUsLinkComponent
Hydrogen peroxide 33% unstabilised, TECHNICAL

Hydrogen peroxide 33% unstabilised, TECHNICAL

Plastic drum, 20 L
Catalog # 23613.446
Corrosive
Irritant
Supplier:   VWR Chemicals
Hydrogen peroxide 33% unstabilised, TECHNICAL
Catalog # 23613.446
Supplier:  VWR Chemicals
CAS Number:  7722-84-1
Hydrogen peroxide 33% unstabilised, TECHNICAL
Hydrogen peroxide 33% unstabilised, TECHNICAL

Plastic drum, 20 L

CAS Number:  7722-84-1
Price
464,00 €
Log in for availability
1 * 20 L
Product Details & Documents
  • CAS Number:7722-84-1
  • Formula:H₂O₂
  • MDL Number:MFCD00011333
  • Molecular Weight:34.01 g/mol
  • Boiling Pt:107 °C (1013 hPa)
  • Melting Pt:–26 °C
  • Density:1.11 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-765-0
  • UN:2014
  • ADR:5.1 (8),II
  •  :Density: 1.11 g/cm³ (20 °C)
Specifications

Specification Test Results

  • Assay:
    31 - 36 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    20 L

Specifications