Hydrogen peroxide 33% unstabilised, TECHNICAL
Plastic bottle, 5 L
Supplier: VWR Chemicals
Hydrogen peroxide 33% unstabilised, TECHNICAL
Catalog # 23613.366
Supplier: VWR Chemicals
CAS Number: 7722-84-1
Jump to:
Product Details & Documents
- CAS Number:7722-84-1
- Formula:H₂O₂
- MDL Number:MFCD00011333
- Molecular Weight:34.01 g/mol
- Boiling Pt:107 °C (1013 hPa)
- Melting Pt:–26 °C
- Density:1.11 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-765-0
- UN:2014
- ADR:5.1 (8),II
- :Density: 1.11 g/cm³ (20 °C)
Documents
Certificates
Search by Lot Number1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500) 5L Plastic bottle for liquids DIN 45 UN X (ZPACK13951035)
Packaging Information
P20_VWRC23613.297_SPEC_EN.pdf P20_VWRC23613.366_SPEC_EN.pdf spec_VWRC23613.297 spec_VWRC23613.366 spec_VWRC23613.446
Product specification
More
VWR Chemicals Big Packs and Bulks
Product Literature
Specifications
Specification Test Results
- Assay:31 - 36 %
Specifications
- Pack type:Plastic bottle
- Pk:5 L



