Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®

Metal drum, 25 L
Catalog # 23532.468
Flammable
Health
Irritant
Supplier:   VWR Chemicals
1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®
Catalog # 23532.468
Supplier:  VWR Chemicals
CAS Number:  123-91-1
1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®
1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®

Metal drum, 25 L

CAS Number:  123-91-1
Price
1 260,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
  • CAS Number:123-91-1
  • Formula:C₄H₈O₂
  • MDL Number:MFCD00006571
  • Molecular Weight:88.11 g/mol
  • Merck Index:13,03330
  • Boiling Pt:101.1 °C (1013 hPa)
  • Melting Pt:11.8 °C
  • Density:1.03 g/cm³ (20 °C)
  • Flash Pt:12 °C
  • Storage Temperature:Ambient
  • UN:1165
  • ADR:3,II
  •  :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
  • EINECS:204-661-8
Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Boiling point:
    99 - 102 °C
  • Density (20/4):
    1.030 - 1.035

Specifications

  • Pack type:
    Metal drum
  • Pk:
    25 L

Specifications