1,4-Dioxane ≥99% stabilised, GPR RECTAPUR®
Glass bottle, 1 L
: Glycolethylether, 1,4-Dioxacyclohexane, Dioxane, p-Dioxane, 1,4-Diethylene dioxide
- :123-91-1
- :C₄H₈O₂
- :MFCD00006571
- :88.11 g/mol
- :13,03330
- :101.1 °C (1013 hPa)
- :11.8 °C
- :1.03 g/cm³ (20 °C)
- :12 °C
- :Ambient
- :1165
- :3,II
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 25 ppm
- :204-661-8
- Assay:Min. 99 %
- Boiling point:99 - 102 °C
- Density (20/4):1.030 - 1.035
- Pack type:Glass bottle
- Pk:1 L



