Order Entry
Export
Orders LinkContactUsLinkComponent
Ion selective electrodes, cupric, Orion™

Ion selective electrodes, cupric, Orion™

Cupric solid-state half-cell, 9429BN, 1×10-8 - 0,1 M (6,4×10-4 - 6350 ppm), 0...+80 °C, 9429BN, BNC, 1 m cable
Catalog # 662-0841
Supplier:   Thermo Orion
Ion selective electrodes, cupric, Orion™
Catalog # 662-0841
Supplier:  Thermo Orion
Ion selective electrodes, cupric, Orion™

Cupric solid-state half-cell, 9429BN, 1×10-8 - 0,1 M (6,4×10-4 - 6350 ppm), 0...+80 °C, 9429BN, BNC, 1 m cable

Price
939,00 €
Log in for availability
1 * 1 items
Product Details & Documents

These electrodes are designed for measuring cupric ions in aqueous solutions quickly, simply, accurately and economically.

  • Stable, drift-free potentials
  • Sure-Flow™ junction allows easy cleaning and is easy to use in tough samples
  • When conventional electrode junctions clog, the Sure-Flow™ junction can be opened and flushed clean simply by pressing the cap
  • Half-cell electrode used in conjunction with the double junction reference electrode offers the benefits of the Sure-Flow™ junction design
  • Combination electrode ORIO9629BNWP does not require a separate reference electrode, making it convenient to use with small sample volumes

Ordering information: The half cell electrodes require a separate reference electrode which can be ordered separately.

Specifications
  • Concentration:
    1×10-8 - 0,1 M (6,4×10-4 - 6350 ppm)
  • Connection:
    BNC, 1 m cable
  • Type:
    9429BN
  • Description:
    Cupric solid-state half-cell, 9429BN
  • Temp.:
    0...+80 °C
Accessories

Product Information

  • Catalog #
    Symbol
    Concentration
    Matrix
    Availability
    Price
  • 662-0840
    Cu
    0,1M
    H₂O
    Log in for availability
    1 * 475 mL - 141,00 €

Product Information

  • Catalog #
    Type
    Pack type
    Availability
    Price
  • Ionic strength adjuster for chloride ion selective electrode, in a bottle
    Ionic strength adjuster for chloride ion selective electrode, in a bottle
    662-0824
    Ion selective electrode reagents
    Bottle
    Log in for availability
    1 * 475 mL - 102,00 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    Potassium nitrate electrode filling solution for double junction Sure-Flow reference pH electrodes, outer chamber, 60 ml bottle
    662-1524
    5
    Log in for availability
    1 * 5 items - 121,00 €

Product Information

  • Catalog #
    Pk
    Availability
    Price
  • Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    Potassium chloride & silver chloride electrode filling solution for double junction Sure-Flow reference pH electrodes, inner chamber, 60 ml bottle
    662-0750
    5
    Log in for availability
    1 * 5 items - 121,00 €