Order Entry
Export
Orders LinkContactUsLinkComponent
 

Passive gas detectors, Dositubes

Chlorine (8D), Chlorine, 0,08 - 100 ppm
  151-0091
 :   GASTEC
Passive gas detectors, Dositubes
  151-0091
 :  GASTEC
Passive gas detectors, Dositubes
Passive gas detectors, Dositubes

Chlorine (8D), Chlorine, 0,08 - 100 ppm

 
62,70 €
   
1 * 10 items
 

The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.

  • Compact, lightweight, and versatile (can be used anywhere)
  • Easy direct measuring and reading
  • No sampling pump necessary

The Dositubes use the principle of the natural diffusion of gas.

A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.

 
  • Description:
    Chlorine (8D)
  • Parameter(s):
    Chlorine
  • Range:
    0,08 - 100 ppm
  • Cat. No.:
    151-0091
  • Measurement time:
    0,5 - 24 h
Accessories

 

  •  
    Description
    Parameter(s)
    Range
     
     
  • 151-0090
    Acetic acid (81D)
    Acetic acid
    0,5 - 100 ppm
       
    1 * 10 items - 62,70 €

 

  •  
    Description
     
     
  • 151-0095
    Holder for Dositube
       
    1 * 3 items - 47,00 €