Passive gas detectors, Dositubes
Perchlorethylene (133D), Perchlorethylene, 3 - 300 ppm
Catalog # 151-0075
Supplier: GASTEC
Jump to:
Product Details & Documents
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
- Compact, lightweight, and versatile (can be used anywhere)
- Easy direct measuring and reading
- No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
Specifications
- Description:Perchlorethylene (133D)
- Parameter(s):Perchlorethylene
- Range:3 - 300 ppm
- Cat. No.:151-0075
- Measurement time:1 - 8 h
Accessories
Product Information
- Catalog #Parameter(s)RangeAvailabilityPrice
- Acetic acid (81D)
Product Information
- Catalog #AvailabilityPrice
- Holder for Dositube



