Order Entry
Export
Orders LinkContactUsLinkComponent
Passive gas detectors, Dositubes

Passive gas detectors, Dositubes

Hydrogen chloride (14D), Hydrogen chloride, 1 - 100 ppm
Catalog # 151-0076
Supplier:   GASTEC
Passive gas detectors, Dositubes
Catalog # 151-0076
Supplier:  GASTEC
Passive gas detectors, Dositubes
Passive gas detectors, Dositubes

Hydrogen chloride (14D), Hydrogen chloride, 1 - 100 ppm

Price
62,70 €
Log in for availability
1 * 10 items
Product Details & Documents

The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.

  • Compact, lightweight, and versatile (can be used anywhere)
  • Easy direct measuring and reading
  • No sampling pump necessary

The Dositubes use the principle of the natural diffusion of gas.

A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.

Specifications
  • Description:
    Hydrogen chloride (14D)
  • Parameter(s):
    Hydrogen chloride
  • Range:
    1 - 100 ppm
  • Cat. No.:
    151-0076
  • Measurement time:
    1 - 8 h
Accessories

Product Information

  • Catalog #
    Parameter(s)
    Range
    Availability
    Price
  • Acetic acid (81D)
    Acetic acid (81D)
    151-0090
    Acetic acid
    0,5 - 100 ppm
    Log in for availability
    1 * 10 items - 62,70 €

Product Information

  • Catalog #
    Availability
    Price
  • Holder for Dositube
    Holder for Dositube
    151-0095
    Log in for availability
    1 * 3 items - 47,00 €