Passive gas detectors, Dositubes
Acetic acid (81D), Acetic acid, 0,5 - 100 ppm
151-0090
: GASTEC
The dositubes are used to measure time-weighted average (TWA) gas concentrations over longer periods.
- Compact, lightweight, and versatile (can be used anywhere)
- Easy direct measuring and reading
- No sampling pump necessary
The Dositubes use the principle of the natural diffusion of gas.
A Dositube is a glass tube of a specific inner diameter that is filled with a diffuser and chemical reagent in parallel that enable gas to disperse at a constant rate resulting in a longer discolouration and better demarcation. The passive Dositube can then be positioned in the worker's breathing zone where it can be attached with a holder and collar clip.
- Description:Acetic acid (81D)
- Parameter(s):Acetic acid
- Range:0,5 - 100 ppm
- Cat. No.:151-0090
- Measurement time:1 - 10 h



