Orders LinkContactUsLinkComponent
VWR® Labomeca®, Feet for rods
16 mm Ø rods
  241-0136
VWR® Labomeca®, Feet for rods
  241-0136
VWR® Labomeca®, Feet for rods
VWR® Labomeca®, Feet for rods
16 mm Ø rods
201,00 €
1 * 10 items
Mounting brackets for Labomeca® assemblies.
  • Used for attaching rods to a fixed point of the laboratory: Floor, walls, ceiling, tables, shelves and furniture
  • Supplied with wood screw, can be secured by using Labomeca® Allen key
 : Supplied with wood screw.
  • For:
    16 mm Ø rods