Millex® Polyethersulfone Inline Filters
Inline filter, 6,35 - 12,7 mm stepped hose barb with Female Luer slip interior, outlet fitting bell, 0,22 μm, 19,6 cm², 50 mm, +
Catalog # MILFSLGPB5010
Supplier: Merck
Jump to:
Product Details & Documents
The bi-directionally supported Millex-GP50 filter is a sterilising filter for aqueous solutions. It removes microorganisms, particles, precipitates, and undissolved powders larger than 0,22 μm. This single-use product consists of a 0,22 μm membrane filter sealed in an acrylic housing.
- Sterile filtration of tissue culture media, additives, buffers, and water
- Sterile, gamma irradiated
- Hydrophilic
- Millex-GP50 filter is non-pyrogenic and non-toxic
For use with peristaltic pump, a pump drive and pump head that delivers a constant speed of 60 revolutions per minute (rpm) is recommended. This generates an average flow rate of 100 ml/min. Recommended tubing: 4 to 9,5 mm Int. Ø. silicone rubber or latex tubing.
Documents
Cobranded Syringe Filters.pdf Merck Syringe Filters.pdf
Product Literature
Specifications
- Ø:50 mm
- Filtration area:19,6 cm²
- Pore size:0,22 μm
- Sterile:+
- Description:Inline filter, 6,35 - 12,7 mm stepped hose barb with Female Luer slip interior, outlet fitting bell
- Seq:02
- Cat. No.:MILFSLGPB5010
- Max. pressure:3,3 bar
- Housing material:Methacrylate-butadiene-styrene (MBS)
- Inlet/outlet connection:96 mm, including bell



