Order Entry
Export
Orders LinkContactUsLinkComponent
Millex® Polyethersulfone Inline Filters

Millex® Polyethersulfone Inline Filters

Inline filter, 6,35 - 12,7 mm hose barb with Female Luer slip interior, 0,22 μm, 19,6 cm², 50 mm, +
Catalog # MILFSLGP05010
Supplier:   Merck
Millex® Polyethersulfone Inline Filters
Catalog # MILFSLGP05010
Supplier:  Merck
Millex® Polyethersulfone Inline Filters
Millex® Polyethersulfone Inline Filters

Inline filter, 6,35 - 12,7 mm hose barb with Female Luer slip interior, 0,22 μm, 19,6 cm², 50 mm, +

Price
199,00 €
Log in for availability
1 * 10 items
Product Details & Documents

The bi-directionally supported Millex-GP50 filter is a sterilising filter for aqueous solutions. It removes microorganisms, particles, precipitates, and undissolved powders larger than 0,22 μm. This single-use product consists of a 0,22 μm membrane filter sealed in an acrylic housing.

  • Sterile filtration of tissue culture media, additives, buffers, and water
  • Sterile, gamma irradiated
  • Hydrophilic
  • Millex-GP50 filter is non-pyrogenic and non-toxic

For use with peristaltic pump, a pump drive and pump head that delivers a constant speed of 60 revolutions per minute (rpm) is recommended. This generates an average flow rate of 100 ml/min. Recommended tubing: 4 to 9,5 mm Int. Ø. silicone rubber or latex tubing.

Specifications
  • Ø:
    50 mm
  • Filtration area:
    19,6 cm²
  • Pore size:
    0,22 μm
  • Sterile:
    +
  • Description:
    Inline filter, 6,35 - 12,7 mm hose barb with Female Luer slip interior
  • Seq:
    01
  • Cat. No.:
    MILFSLGP05010
  • Max. pressure:
    3,3 bar
  • Housing material:
    Methacrylate-butadiene-styrene (MBS)
  • Inlet/outlet connection:
    96 mm, including bell