Digital Solutions
Applications
Support
About
Export
Orders Link
ContactUsLinkComponent
Retort Rods
VWR® Retort Rods, Unthreaded
VWR® Retort Rods, Unthreaded
16 mm, 500 mm
241-0085
:
VWR Collection
VWR® Retort Rods, Unthreaded
241-0085
:
VWR Collection
VWR® Retort Rods, Unthreaded
16 mm, 500 mm
45,50 €
1 * 1 items
-
+
Stainless steel, hollow.
Excellent chemical resistance
Durable hollow rods reducing weight in support constructions
VWR® for filtration
VWR® for Stirring, Mixing, Shaking And Blending
Ø:
16 mm
Length:
500 mm