Filter bags, BagPage®
Filter bag 3500 with filter porosity: 280 microns, optimal blending volume 200 − 3500 ml, 3500 ml, 10/pack, 100/box, 600×380 mm
Supplier: INTERSCIENCE
Jump to:
Product Details & Documents
BagPage® single use bags with filter membrane reduce analysis time thanks to their wide opening and markings showing where to introduce the pipette. Ideal for pasty products, cheese, flour and cakes. To use: Place the sample in the opened compartment (Fig. 1). Mix and then add reagents into the specially designed compartment (Fig. 2).
- Strong with wide opening for introducing the sample and easy pipetting
- Pipettes are not blocked by food fragments, which are retained behind the filter membrane
- Transparent and rigid with a full page micro perforated, calibrated filter
- Sterilised by irradiation, supplied with sterilisation certificate
Specifications
- Packed:10/pack, 100/box
- Capacity:3500 ml
- Description:Filter bag 3500 with filter porosity: 280 microns, optimal blending volume 200 − 3500 ml
- Seq:09
- W×L:600×380 mm



