Bottles, serum, WHEATON®
60,00 ml, 38×84 mm, Natural, No, 20 mm
WHEA224036
These bottles are manufactured from 400 borosilicate glass that conforms to USP Type I requirements.
- Accepts standard stoppers and aluminum seals
- PP bottle is used extensively for diluent solutions
- HDPE bottle is widely used for animal health products and storage of non-medical products
- Autoclavable at 121 °C for 20 minutes
- Cap included:No
- Cap Size:20 mm
- Ø×H:38×84 mm
- Capacity:60,00 ml
- Colour:Natural
- Seq:06



