Order Entry
Export
Orders LinkContactUsLinkComponent
Bottles, serum, WHEATON®

Bottles, serum, WHEATON®

30,00 ml, 32×67 mm, Natural, No, 20 mm
Catalog # WHEA224033
Supplier:   DWK Life Sciences
Bottles, serum, WHEATON®
Catalog # WHEA224033
Supplier:  DWK Life Sciences
Bottles, serum, WHEATON®
Bottles, serum, WHEATON®

30,00 ml, 32×67 mm, Natural, No, 20 mm

Price
476,00 €
Log in for availability
1 * 200 items
Product Details & Documents

These bottles are manufactured from 400 borosilicate glass that conforms to USP Type I requirements.

  • Accepts standard stoppers and aluminum seals
  • PP bottle is used extensively for diluent solutions
  • HDPE bottle is widely used for animal health products and storage of non-medical products
  • Autoclavable at 121 °C for 20 minutes
Specifications
  • Cap included:
    No
  • Cap Size:
    20 mm
  • Ø×H:
    32×67 mm
  • Capacity:
    30,00 ml
  • Colour:
    Natural
  • Seq:
    05