Bottles, serum, WHEATON®
15,00 ml, 28×56 mm, Natural, No, 20 mm
Catalog # WHEA224031
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These bottles are manufactured from 400 borosilicate glass that conforms to USP Type I requirements.
- Accepts standard stoppers and aluminum seals
- PP bottle is used extensively for diluent solutions
- HDPE bottle is widely used for animal health products and storage of non-medical products
- Autoclavable at 121 °C for 20 minutes
Specifications
- Cap included:No
- Cap Size:20 mm
- Ø×H:28×56 mm
- Capacity:15,00 ml
- Colour:Natural
- Seq:04



