Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Closures
Bottle Closures
Screw caps, PE
Screw caps, PE
Print
Screw caps, PE
self-sealing, 40 mm, Natural, PE
Catalog # 215-2694
Supplier:
GRAVIS
Screw caps, PE
Catalog # 215-2694
Supplier:
GRAVIS
Expand image
Return to Top
Screw caps, PE
self-sealing, 40 mm, Natural, PE
Specifications
Add to cart
Jump to:
Specifications
Price
61,40 €
Log in
for availability
Unit of Measure
1 * 100 items
-
+
Add to cart
Other Product Family Options Available
Return Policy
Product Details & Documents
Specifications
Thread:
40 mm
Colour:
Natural
Description:
self-sealing
Material:
PE