Orders LinkContactUsLinkComponent
Cables for Screw Cap Electrodes
Cables for Screw Cap Electrodes
Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument sockets, 1 m
Catalog # HACHA94L114
Supplier:   Hach
Cables for Screw Cap Electrodes
Catalog # HACHA94L114
Supplier:  Hach
Cables for Screw Cap Electrodes
Cables for Screw Cap Electrodes
Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument sockets, 1 m
Price
108,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Electrode connection cable, CL114, for connecting screw cap electrodes (S7) to BNC instrument sockets
  • Length:
    1 m
  • Seq:
    02