Order Entry
Export
Orders LinkContactUsLinkComponent
Trolley, trays

Trolley, trays

Trays trolley, with guard rails, two handles and three trays, argon welded, 600W×900L mm, 3
Catalog # AGEN947S
Supplier:   Agencinox
Trolley, trays
Catalog # AGEN947S
Supplier:  Agencinox
Trolley, trays
Trolley, trays

Trays trolley, with guard rails, two handles and three trays, argon welded, 600W×900L mm, 3

Price
1 970,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.

  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models
Specifications
  • Description:
    Trays trolley, with guard rails, two handles and three trays, argon welded, 600W×900L mm
  • Seq:
    10
  • Shelves:
    3