Order Entry
Export
Orders LinkContactUsLinkComponent
 

Trolley, trays

Trays trolley, without guard rails, two handles and two trays, knockdown construction, 430W×600L mm, 2
Catalog # 139-0100
Supplier:   Agencinox
Trolley, trays
Catalog # 139-0100
Supplier:  Agencinox
Trolley, trays
Trolley, trays

Trays trolley, without guard rails, two handles and two trays, knockdown construction, 430W×600L mm, 2

Price
573,00 €
Log in for availability
1 * 1 items
Product Details & Documents

Trolleys with two handles, shaped trays made up of 18/10 stainless steel with bright polish finishing.

  • Heavy duty structure
  • Easy cleaning
  • Available in knockdown construction or argon welded models
Specifications
  • Description:
    Trays trolley, without guard rails, two handles and two trays, knockdown construction, 430W×600L mm
  • Seq:
    01
  • Shelves:
    2