Order Entry
Export
Orders LinkContactUsLinkComponent
Water, GPR RECTAPUR®

Water, GPR RECTAPUR®

Bag-in-box, 10 L
Catalog # 83612.400
Supplier:   VWR Chemicals
Water, GPR RECTAPUR®
Catalog # 83612.400
Supplier:  VWR Chemicals
CAS Number:  7732-18-5
Water, GPR RECTAPUR®
Water, GPR RECTAPUR®

Bag-in-box, 10 L

CAS Number:  7732-18-5
Price
84,30 €
Log in for availability
1 * 10 L
Product Details & Documents
Synonyms:  Hydrogen oxide
  • CAS Number:7732-18-5
  • Formula:H₂O
  • MDL Number:MFCD00011332
  • Molecular Weight:18.02 g/mol
  • Merck Index:13,10098
  • Boiling Pt:100 °C (1013 hPa)
  • Melting Pt:0 °C
  • Density:1 g/cm³ (4 °C)
  • Storage Temperature:Ambient
  • EINECS:231-791-2
Specifications

Specification Test Results

  • Acidity or alkalinity:
    Passes test
  • Oxidisable substances:
    Passes test
  • Conductivity (20°C):
    Max. 1.5 µS/cm
  • Evaporation residue:
    Max. 1 ppm
  • Heavy metals (as Pb):
    Max. 0.1 ppm
  • Cl (Chloride):
    Max. 0.1 ppm
  • NH4 (Ammonium):
    Max. 0.2 ppm
  • NO3 (Nitrate):
    Max. 0.2 ppm
  • SO4 (Sulphate):
    Max. 1 ppm
  • Ca (Calcium):
    Max. 0.1 ppm
  • Na (Sodium):
    Max. 0.1 ppm

Specifications

  • Pack type:
    Bag-in-box
  • Pk:
    10 L

Specifications