Order Entry
Export
Orders LinkContactUsLinkComponent
 

Water, GPR RECTAPUR®

Plastic drum, 25 L
  83612.460
Water, GPR RECTAPUR®
  83612.460
 :  7732-18-5
Water, GPR RECTAPUR®
Water, GPR RECTAPUR®

Plastic drum, 25 L

 :  7732-18-5
 
127,00 €
   
1 * 25 L
 

 

  • Acidity or alkalinity:
    Passes test
  • Oxidisable substances:
    Passes test
  • Conductivity (20°C):
    Max. 1.5 µS/cm
  • Evaporation residue:
    Max. 1 ppm
  • Heavy metals (as Pb):
    Max. 0.1 ppm
  • Cl (Chloride):
    Max. 0.1 ppm
  • NH4 (Ammonium):
    Max. 0.2 ppm
  • NO3 (Nitrate):
    Max. 0.2 ppm
  • SO4 (Sulphate):
    Max. 1 ppm
  • Ca (Calcium):
    Max. 0.1 ppm
  • Na (Sodium):
    Max. 0.1 ppm

 

  • Pack type:
    Plastic drum
  • Pk:
    25 L