Water, GPR RECTAPUR®
Plastic container, 5 L
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Hydrogen oxide
- CAS Number:7732-18-5
- Formula:H₂O
- MDL Number:MFCD00011332
- Molecular Weight:18.02 g/mol
- Merck Index:13,10098
- Boiling Pt:100 °C (1013 hPa)
- Melting Pt:0 °C
- Density:1 g/cm³ (4 °C)
- Storage Temperature:Ambient
- EINECS:231-791-2
Certificates
Search by Lot Number10L Cubitainer UN (ZPACK13260004) 10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Specifications
Specification Test Results
- Acidity or alkalinity:Passes test
- Oxidisable substances:Passes test
- Conductivity (20°C):Max. 1.5 µS/cm
- Evaporation residue:Max. 1 ppm
- Heavy metals (as Pb):Max. 0.1 ppm
- Cl (Chloride):Max. 0.1 ppm
- NH4 (Ammonium):Max. 0.2 ppm
- NO3 (Nitrate):Max. 0.2 ppm
- SO4 (Sulphate):Max. 1 ppm
- Ca (Calcium):Max. 0.1 ppm
- Na (Sodium):Max. 0.1 ppm
Specifications
- Pack type:Plastic container
- Pk:5 L



