Safety spectacles, Op-Tema®
Anti-scratch, Black, Clear PC
Catalog # 111-3500
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
Comfortable safety spectacles with lightweight full view lens and built-in moulded browbar.
- Soft, adjustable nose piece
- Fully adjustable
- Adjustable, break-away Flexicord retainer for added safety
Weight: 28 g
: EN 166, EN 170; 1F
Specifications
- Type:Anti-scratch
- Colour:Black
- Seq:01
- Lenses:Clear PC