Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Rectangular comb for 33cm wide units. 0.5mm x 63 well formatted for us with multi 1 * 1 items
Catalog # CBSCSG33-0563
Supplier:
Expedeon
Expand image
Rectangular comb for 33cm wide units. 0.5mm x 63 well formatted for us with multi 1 * 1 items
Catalog # CBSCSG33-0563
Supplier:
Expedeon