Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Comb for mini-vertical unit 11.3cm width. 3.0mm x 7 well 1 * 1 items
CBSCMVC-3007
:
Expedeon
Comb for mini-vertical unit 11.3cm width. 3.0mm x 7 well 1 * 1 items
CBSCMVC-3007
:
Expedeon