Orders LinkContactUsLinkComponent
Distilling receivers, Bernauer
14/23 NS
  LENZ05288001
Distilling receivers, Bernauer
  LENZ05288001
Distilling receivers, Bernauer
Distilling receivers, Bernauer
14/23 NS
118,00 €
1 * 1 items
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Documents
  • Socket size:
    14/23 NS