Erlenmeyer flasks, narrow neck, with baffles
2000 ml
Catalog # NEUB0749-30-2000
Supplier: NEUBERT VOLUME GLASSWAERE
Erlenmeyer flasks, narrow neck, with baffles
Catalog # NEUB0749-30-2000
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.
Specifications
- Capacity:2000 ml
- Seq:11