Orders LinkContactUsLinkComponent
Erlenmeyer flasks, narrow neck, with baffles
500 ml
Catalog # 214-0186
Erlenmeyer flasks, narrow neck, with baffles
Catalog # 214-0186
Erlenmeyer flasks, narrow neck, with baffles
500 ml
Price
66,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.
Specifications
  • Capacity:
    500 ml
  • Seq:
    09