Nessler cylinders
Nessler cylinders, 50 ml, tall form, Ø×H: 21×265, 50 ml division
Catalog # 214-0179
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
- Optically polished, flat base
- With pouring spout
Specifications
- Description:Nessler cylinders, 50 ml, tall form, Ø×H: 21×265, 50 ml division
- Seq:01