Accessories for Corning® spinner flasks
Dual ¹/₄" media handling fitting, for 8 L and 15 L flasks
Catalog # 734-1945
Supplier: Corning
Jump to:
Product Details & Documents
Documents
Specifications
- Description:Dual ¹/₄" media handling fitting, for 8 L and 15 L flasks
- Seq:18



