ContactUsLinkComponent
Microplate 1536 well polystyrene Low Volume 2µl clear flat bottom High Bind without lid non-sterile 1 * 100 items
Catalog # 734-1656
Supplier:  Corning
Microplate 1536 well polystyrene Low Volume 2µl clear flat bottom High Bind without lid non-sterile 1 * 100 items
Catalog # 734-1656
Supplier:  Corning